Order Entry
Sweden
ContactUsLinkComponent
 
Anti-ST6GAL1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3793R
Supplier:  Bioss
undefined
Anti-ST6GAL1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3793R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ST6 beta-galactosamide alpha-2,6-sialyltranferase 1
  • Antigen symbol:
    ST6GAL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Antigen synonyms:
    B-cell antigen CD75|6-sialyltransferase 1|alpha 2|ST6GalI|6-sialyltransferase|ST6Gal I|CMP-N-acetylneuraminate beta-galactosamide alpha-2|6-ST 1|ST6N|beta-galactoside alpha-2|sialyltransferase 1|CMP-N-acetylneuraminate-beta-galactosamide-alpha-2|Sialyltransferase 1|SIAT1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: ST6GAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat