Anti-Nur77 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3513R
Leverantör: Bioss
Specifikationer
- Antibody type:Primary
- Antigen name:Nuclear Receptor Subfamily 4 Group A Member 1
- Antigen symbol:NR4A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:Orphan nuclear receptor HMR|HMR|ST-59|Orphan nuclear receptor NR4A1|GFRP1|NR4A1|NAK1|Early response protein NAK1|TR3 orphan receptor
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: Nur77
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat