Anti-HSD3B7 Rabbit Polyclonal Antibody
Catalog # BOSSBS-2366R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Hydroxysteroid (3-beta) Dehydrogenase 7
- Antigen symbol:HSD3B7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:C(27)-3BETA-HSD|3-beta-HSD VII|3 beta-hydroxy-delta 5-C27-steroid oxidoreductase|cholest-5-ene-3-be|SDR11E3|3-beta-hydroxy-Delta(5)-C27 steroid oxidoreductase|3 beta-hydroxysteroid dehydrogenase type 7|c(27) 3-beta-HSD|PFIC4|3 beta-hydroxysteroid dehydrogenase type VII
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: HSD3B7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat