Anti-IGFBP3 Rabbit Polyclonal Antibody
BOSSBS-1434R
: Bioss
- Antibody type:Primary
- Antigen name:Insulin-Like Growth Factor Binding Protein 3
- Antigen symbol:IGFBP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: IGFBP3 (insulin-like growth factor binding protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat