Anti-CEACAM8 Rabbit Polyclonal Antibody
BOSSBS-0744R
: Bioss
- Antibody type:Primary
- Antigen name:Carcinoembryonic antigen-related cell adhesion molecule 8
- Antigen symbol:CEACAM8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:CEACAM8|CD66b|NCA-95|CGM6|CD67
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: CEACAM8
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: