Anti-JUN Rabbit Polyclonal Antibody
BOSSBS-0670R
: Bioss
- Antibody type:Primary
- Antigen name:c-Jun
- Antigen symbol:JUN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:Transcription factor AP-1|V-jun avian sarcoma virus 17 oncogene homolog|JUN|AP1|p39|Proto-oncogene c-jun|Activator protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: JUN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat