VWR®, Bottle Carrier
Katalog # 215-3582
Leverantör: VWR Collection
Specifikationer
- Capacity:2,5 l
- Colour:White
- Description:Bottle carrier with safe handle
- Pk:1
Specifikationer
Om denna produkt
Glass bottles are designed to contain liquids, but when they break, hazardous liquids potentially mix with broken glass. This bottle carrier protects people and property from glass shards, whilst limiting exposure to flammable or corrosive liquids.
- Minimises spills and breakage
- Easy open/close mechanism
- Environmentally friendly: Durable and liquid resistant handle construction