Industrial gloves, oil repellent, HyFlex® 11-926
Supplier: Ansell
Certificates
About this item
This knitted Spandex/nylon glove features a ¾ dipped neoprene/nitrile coating which provides oil repellence. Ideal for use in oily environments.
- RIPEL™ technology creates a liquid-impermeable layer that prevents oil or lubricant from making even incidental contact with the wearer’s skin
- Durable ¾ dipped coating ensures full protection for the back of the fingers and a safe grip in oily environments
- Liner is made with one of the finest gauge nylon fibre
- Enhanced flexibility and tactility
Suitable applications include: Stamping light and small metal parts, primary assembly of light components, maintenance operations, loading and downloading parts in metal working lines and assembly in machining operations.
Certifications: EN 388:2016 (4121A); Cat. II.
Specifications
- Catalog No:
- 112-4291
- 112-4294
- 112-4290
- 112-4295
- 112-4293
- 112-4292
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Spandex/nylon
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)