Filter boards
Supplier: Sartorius
Certificates
About this item
Filter boards are used for filtration of cooking oils, transformer oils and as base paper for impregnation with certain reagents.
These are characterised by high wet strength and are therefore especially suitable for pressure or vacuum filtration.
Specifications
- Catalog No:
- SARTFT-3-343-293
- SARTFT-3-343-142
- SARTFT-3-343-047
- Grade:
- C 160
- C 160
- C 160
- Weight:
- 160 g/m²
- 160 g/m²
- 160 g/m²
- Thickness:
- 0,30 mm
- 0,30 mm
- 0,30 mm
- Filtration speed:
- -
- -
- -
- Wet burst strength:
- >50 kPa
- >50 kPa
- >50 kPa
- Capillary rise:
- 80 mm/10 min
- 80 mm/10 min
- 80 mm/10 min