HPLC columns, MAbPac™ HIC-Butyl
The MAbPac™ HIC-Butyl HPLC columns are used to separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs). The column's hydrophilic resin provides excellent biocompatibility and low carryover, while the optimal density of the butyl groups provides high resolution separation.
- High resolution and high efficiency
- Stable at wide range of pH (2 to 12)
- Low carryover
- Rugged column packing
- Catalog No:
- 555-7361
- 555-7360
- Max. pressure:
- 4000 psi
- 4000 psi
- pH range:
- 2 - 12
- 2 - 12
- Temperature limit:
- 60 °C
- 60 °C