Heat sealer, Audion 300 MHS
Catalog # 710-1665
Supplier: Local
Specifications
- Description:Heat sealer, Audion 300 MHS
- Pk:1
Specifications
About this item
Compact table top medical heat sealer for sterile and hermetic packaging of medical tools.
- Sealer ready for use in 4 seconds, when the machine gives a light and acoustic signal
- Integrated work table gives extra comfort while working
- Able to accommodate a large selection of roll widths
- Fixed film reel holder makes it possible to use different roll sizes at the same time
- Cutting device enables the operator to create the required length of package
- Heated top seal bar has a fixed temperature and gives a complete seal
IP 20 Class 1; EN ISO 12100, EN IEC 60204-1, EN 1037+A1; 2006/95/EC, 2004-108-EC