Rabbit serum
Supplier: Rockland Immunochemicals
Certificates
About this item
Sera is obtained from non-hemolyzed blood collected from healthy, fasted donors. Animals are maintained on an antibiotic-free diet.
- Available sterile filtered or non sterile
- Concentration of 65,0 mg/ml by refractometry
- Store container at −20 °C prior to opening