Tubing, chloroprene
Chloroprene, black.
- Resistance to hydrocarbons and mineral oils
- Good ageing: ozone resistant
- High di-electrical strength
With its good mechanical, chemical, electrical and heat resistance (thermal) properties, chloroprene is recommended for a wide range of industrial applications. It is used as much for cable insulation, as for transfer of nitrogen and helium gases. It shows an outstanding resistance to UV and weathering.
Temperature resistance: −20 to +100 °C
Hardness (Shore durometer A): 50
Density: 1,55