Order Entry
Sweden
ContactUsLinkComponent
Analytical guard column, Upchurch Scientific®
Analytical guard column, Upchurch Scientific®
Catalog # UPCHC-130B
undefined
Analytical guard column, Upchurch Scientific®
Catalog # UPCHC-130B
Specifications
  • Column Format:
    Guard column
  • Ø int.:
    2,00 mm
  • Description:
    C-130B guard column including 2×A-100
  • Length:
    20 mm
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
This narrow-bore short column (2,0×20 mm) creates only a slight pressure increase with virtually no detectable theoretical plate loss when used with a 3 mm int.Ø or larger column.
  • Available as a kit with funnel and extra frits
  • Easy to pack