Order Entry
Sweden
ContactUsLinkComponent
96-well qPCR strip tube plate, VersiPlate
Catalog # 732-2590
undefined
96-well qPCR strip tube plate, VersiPlate
Catalog # 732-2590
Specifications
  • No. of wells:
    96
  • Profile:
    Low
  • Skirt:
    Non-skirted
  • Well colour:
    White
  • Pk:
    25
  • Well volume:
    0,1 ml
Specifications
About this item
VersiPlates comprise strips of eight tubes linked to each other forming the familiar 12×8 or 96-well ANSI/SBS format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
  • White colouration improves signal reflection, reduces variation and increases the sensitivity of qPCR assays
  • Easier sample handling as multiple strips of samples can be handled simultaneously
  • Reduced consumables waste as only the required numbers of strips need to be removed with the remainder being stored for the next experiment
  • Can be used in instruments with or without a VersiFrame
  • Reusable PC frame can serve as a useful rack for a VersiPlate or any number of low profile strip tubes and facilitates sample loading into thermal cyclers
  • The low profile format of the VersiPlate lessens dead volume, which reduces sample evaporation
VersiCap™ mats
Product Information