Tubings, insulated
Supplier: LAUDA
Certificates
About this item
This insulated tubing is used with thermostats.
*Silicone tubes should not used in conjunction with silicone oils.
This insulated tubing is used with thermostats.
*Silicone tubes should not used in conjunction with silicone oils.