Industrial overalls, Praktika
SSSA02-VPP-XL
- Clothing Size:XL
- Garment application:General Purpose,Industrial
- Gender:Unisex
- Colour:Blue
- Model:Praktika
- Fabric:PP
- Sleeve or leg length:Long
- Weight:50 g/m²
- Pk:100
This versatile overall is made from breathable PP spunbond.
- Washable at 30 °C
- Elastic bands at the sleeves, legs and hood as well as a waist band for adjusting to individual sizes
- Ergonomic, three-part hood
- Cover over zip, to chin
- External overlock seam
- Spacious crotch area with gusset
Perfectly suited for house and garden work, vehicle repairs, construction and paint work, inspections and for protection against hazardous coarse dusts. Can also be used as an overall for visitors.
Colour: blue
Weight: 50 g/m²