Gowning racks, wall-mounted
Supplier: Palbam
Certificates
About this item
Wall-mounted gownracks in stainless steel 304 with electropolished finish.
- Gowning racks include stainless steel hangers
- The design aligns the hangers, keeping an orderly appearance and ensuring the hangers do not touch each other
Delivery: Typically available from stock for direct shipment from the supplier. Shipped flat-packed with assembly instructions.