Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Staining Boxes
Gel staining tray
Gel staining tray
Gel staining tray
Catalog # OWLAGSB-3
Supplier:
THERMO OWL SCIENTIFIC
Expand image
Gel staining tray
Catalog # OWLAGSB-3
Supplier:
THERMO OWL SCIENTIFIC
Specifications
Colour:
Clear
Description:
Staining boxes
W×D×H:
110×110×30 mm
Pk:
5
Specifications
About this item
Product Details
Gel staining tray.
Product Details