Order Entry
Sweden
ContactUsLinkComponent
Gas cylinder holder, Sekuroka®
Gas cylinder holder, Sekuroka®
Catalog # ROTH1513.1
Leverantör:  Roth Carl
Gas cylinder holder, Sekuroka®
Catalog # ROTH1513.1
Leverantör:  Roth Carl
Specifikationer
  • Type:
    Gas cylinder support
  • Pk:
    1
Specifikationer
Om denna produkt
This gas cylinder holder is suitable for safe attachment of gas bottles up to 350 mm Ø. Can be easily fastened onto workbenches, tables, shelves or a ledge up to 60 mm thickness.
  • Bore hole for bolt
  • Variable attachment with black nylon strap