Order Entry
Sweden
ContactUsLinkComponent
Cutter for stainless steel tubing, cutting wheel type
Cutter for stainless steel tubing, cutting wheel type
Katalog # 554-1103
Leverantör:  VICI JOUR RESEARCH
Cutter for stainless steel tubing, cutting wheel type
Katalog # 554-1103
Leverantör:  VICI JOUR RESEARCH
Specifikationer
  • Description:
    Tubing cutter, for cutting 1/16" and 1/8" stainless steel tubing, cutting wheel type
  • Seq:
    01
  • Pk:
    1
Specifikationer
Om denna produkt
This is the ideal tool for cutting ¹/₁₆ and ¹/₈" stainless steel tubing, leaving smooth, uniform cuts requiring a minimum of deburring and/or reaming.
The easily replaced cutting wheel scores tubing, leaving cuts which allow tube-to-tube and tube-to-frit connections.