Chemical resistant overalls, Spacel 3000 AP/RA/EBJ, PE
Leverantör: HONEYWELL SAFETY
Certifikat
Om denna produkt
Liquid tight disposable coverall designed to protect workers from chemicals. Made of three-layered yellow polyethylene film.
- New ergonomics offer a better fitting and protection
- Elasticated wrists and ankles, double adhesive flap and back ventilation valve
- Very good abrasion, tear and puncture resistance
- Watertight due to 'Spaseal' welding
- Yellow colour for improved visibility
Ideal for use in chemical industries, petrochemistry, industrial maintenance, pharmaceutical industry, spoiled beach cleaning, transportation of hazardous materials, industrial sub-contracting cleaning, sanitation, waste processing and emergency services.
Thickness: 100 µm
Certifications: Type 4 + 6; Cat. III.
Specifikationer
- Catalog No:
- DALL4503003-M
- DALL4503003-XL
- DALL4503003-S
- DALL4503003-XXXL
- DALL4503003-XXL
- Model:
- 3000 AP RA EBJ
- 3000 AP RA EBJ
- 3000 AP RA EBJ
- 3000 AP RA EBJ
- 3000 AP RA EBJ
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial