Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrodes
pH Electrodes
Accessories for pH electrodes, InLab® Versatile
Accessories for pH electrodes, InLab® Versatile
Accessories for pH electrodes, InLab® Versatile
Supplier:
Mettler - Toledo
Certificates
Expand image
Accessories for pH electrodes, InLab® Versatile
Supplier:
Mettler - Toledo
About this item