Flocked swabs
Supplier: STERILIN
Certificates
About this item
This innovative nylon fibre technology and unbeatable sample collection and release make the swabs PCR compatible for molecular analysis.
- Manufactured from completely inert components
- Moulded breakpoint that is easily breakable
- Collection vial in various packaging
For forensic purposes product is available certified human DNA free.
Certifications: Certified RNAse- and DNAse-free.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904