Order Entry
Sweden
ContactUsLinkComponent
Sodium sulphate 50 g/l in aqueous solution
Sodium sulphate 50 g/l in aqueous solution
Catalog # FFCHFF63861000
Supplier:  FF CHEMICALS
CAS Number:  
undefined
Sodium sulphate 50 g/l in aqueous solution
Catalog # FFCHFF63861000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
CAS Number:7757-82-6
Formula:Na₂SO₄
MDL Number:MFCD00003504