
2,2,6-Trimethyl-4H-1,3-dioxin-4-one ≥95.0% (by NMR)
Supplier: TCI
Certificates
About this item
CAS Number:5394-63-8
:
Formula:C₇H₁₀O₃
MDL Number:MFCD00040468
:142.15 g/mol
Boiling Pt:77°C
Flash Pt:86°C
EINECS:226-403-3
:GHS07
Specification Test Results
- Appearance:Yellow to Amber to Dark purple clear liquid to cloudy liquid
- Purity (NMR):min. 95.0 %