Zinc chloride ∼7% ∼0.5 mol/L in THF
Catalog # TCIAZ0019-100ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Chelometric Method)unit2:7.0 to 8.5 w/w%
Specification Test Results
About this item
:
:136.3 g/mol
Merck Index:14,10132
EINECS:231-592-0
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
CAS Number:7646-85-7
Formula:ZnCl₂
MDL Number:MFCD00011295