Patton-Reeders reagent (Calconcarboxylic acid) (1:100) diluted with K₂SO₄
Supplier: TCI
Certificates
About this item
CAS Number:3737-95-9
:
Formula:C₂₁H₁₄N₂O₇S
MDL Number:MFCD00004078
:438.42 g/mol
Melting Pt:300 °C
EINECS:223-117-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark red to Dark purple to Dark blue powder to crystaline
- Absorbance:0.600 to 0.800 (Methanol, 550.0 to 560.0 nm)
- Sensitiveness:Colorimetric Test : Turn Purple with Calcium and turn Blue by adding EDTA 2Na