Order Entry
Sweden
ContactUsLinkComponent
 
Bisacrylamide ≥98.0% (by titrimetric analysis) for electrophoresis
 :  TCI
 
undefined
Bisacrylamide ≥98.0% (by titrimetric analysis) for electrophoresis
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Bromination Method):
    min. 98.0 %
  • Functionality test:
    to pass test