
Benzhydrylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5267-34-5
:
Formula:C₁₃H₁₄NCl
MDL Number:MFCD00013023
:219.71 g/mol
Merck Index:14,01076
EINECS:226-084-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Methanol:almost transparency