Order Entry
Sweden
ContactUsLinkComponent
Zinc chloride ∼25% ∼2 mol/L in MeTHF (2-methyltetrahydrofuran)
Zinc chloride ∼25% ∼2 mol/L in MeTHF (2-methyltetrahydrofuran)
Catalog # TCIAZ0020-100ML
Leverantör:  TCI
CAS-nummer:  
undefined
Zinc chloride ∼25% ∼2 mol/L in MeTHF (2-methyltetrahydrofuran)
Catalog # TCIAZ0020-100ML
Leverantör:  TCI
CAS-nummer:  

Specifikationer

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifikationer

Specifikationstestresultat

  • Appearance:
    Colorless to Light yellow clear liquid
  • Concentration (Argentmetric titration):
    1.7 to 2.2 mol/L

Specifikationstestresultat

Om denna produkt

 :
 :136.3 g/mol
EINECS:231-592-0
FN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS08
CAS-nummer:7646-85-7
Formel:ZnCl₂
MDL-nummer:MFCD00011295