Order Entry
Sweden
ContactUsLinkComponent
Zinc chloride 25 mM in aqueous solution
Zinc chloride 25 mM in aqueous solution
Catalog # J62160.AE
CAS Number:  
Zinc chloride 25 mM in aqueous solution
Catalog # J62160.AE
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

CAS Number:7646-85-7
 :
Formula:ZnCl₂
MDL Number:MFCD00011295
 :136.3 g/mol
Storage Temperature:Ambient
EINECS:231-592-0