0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
1-(4-Nitrophenylazo)-2-naphthol
1-(4-Nitrophenylazo)-2-naphthol
1-(4-Nitrophenylazo)-2-naphthol
:
Thermo Fisher Scientific
1-(4-Nitrophenylazo)-2-naphthol
:
Thermo Fisher Scientific
:
6410-10-2
:
:
C₁₆H₁₁N₃O₃
:
MFCD00003908
:
293.28 g/mol
:
248…252 °C
:
229-093-8
:
GHS07
:
Ambient
msds_ALFAB24600
Thermo Scientific Acroseal 25 Years