
1-(4-Nitrophenylazo)-2-naphthol, pure
Catalog # ACRO146300250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Red Powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Elemental Analysis:29.03 to 31.45 % (C)
Specification Test Results
About this item
CAS Number:6410-10-2
:
Formula:C₁₆H₁₁N₃O₃
MDL Number:MFCD00003908
:293.28 g/mol
Melting Pt:248…252 °C
EINECS:229-093-8
:GHS07
Storage Temperature:Ambient