Order Entry
Sweden
ContactUsLinkComponent
 
 
Sodium phenoxide trihydrate for synthesis, Sigma-Aldrich®
 :  Merck
 
 
Sodium phenoxide trihydrate for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    56 - 61 °C
  • Molar Mass:
    170.13 g/mol