Methyl-2-(chloromethyl)benzoate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:34040-62-5
:
Formula:C₉H₉ClO₂
MDL Number:MFCD06200781
:184.6196 g/mol
EINECS:251-805-0
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Light yellow to Amber to Dark green clear liquid
- Purity (GC):min. 97.0 %