Order Entry
Sweden
ContactUsLinkComponent
Thymol blue 0.1% in aqueous solution
Thymol blue 0.1% in aqueous solution
Catalog # FFCHFF2390050
Supplier:  FF CHEMICALS
CAS Number:  
Thymol blue 0.1% in aqueous solution
Catalog # FFCHFF2390050
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :466.6 g/mol
Storage Temperature:Ambient
EINECS:200-973-3
CAS Number:76-61-9
Formula:C₂₇H₃₀O₅S
MDL Number:MFCD00005869