


Fire Assay Flux, Cookson Assay
Catalog # 088-925-01
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:19 kg
- Seq:4238
Specifications
About this item
:
Density:9.96 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2291
ADR:6.1,III
:GHS08
:GHS07
:GHS09