Bunsen burner, Labogaz® 470
Catalog # CAMP2000020657
Supplier: CAMPING GAZ
Specifications
- Description:Bunsen burner, Labogaz® 470
- Seq:01
- Pk:1
- For:Campingaz® valve cartridges CV 270 Plus, CV 300 Plus and CV 470 Plus
Specifications
About this item
Compact bunsen burner for operation with the Campingaz cartridge CV 470 Plus.
- Device can be removed from the cartridge even if the cartridge is not yet completely empty
- Ideal for heating solutions in glass flasks and test tubes or bending parts such as glass tubes
For a wide range of applications in crafts (optics, goldsmith work), industrial laboratories (physics, chemistry, pharmaceuticals) or schools (practical work, experiments).