Human Native Transforming growth factor-β (TGF-β), Corning®
Catalog # BDAA354039
Supplier: Corning
CAS Number:
Specifications
- Pk:1 µG
- Protein/peptide type:Native
- Source:Phlatelets
- Species:Human
- Seq:01
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 14001 - Bedford
- Protein synonyms:Transforming Growth Factor-beta
- Protein/peptide name:TGF-ß
- Purity:Purity > 95% by SDS-PAGE
Specifications
About this item
Transforming Growth Factor-β (TGF-β), human natural, is a multifunctional protein that plays a central role in the regulation of cell growth and differentiation with either stimulatory or inhibitory effects, depending on the context of its action.
Caution: TGF-beta absorbs to glass and plastic surfaces (instructions for dilution are provided with the product).
Tested for ability to inhibit DNA synthesis of Mv 1 Lu mink lung cells (ATCC CCL-64). Tested and found negative for the presence of bacteria, fungi, and mycoplasma.