Drum sumps, large volumes, GW
These drum sumps made of 3 mm fire galvanised sheet steel are used for storing and filling off water-endangering and flammable liquids.
- Removable fire-galvanised grid
- Ground clearance for fork-lift transport
- Models GW 3, GW 5 and GW 6 suitable for storing 1000 L containers
: Models GW 4, GW 5 and GW 6 are supplied as single sump pallets, the single sump pallet are connected together on site with sump pallet connectors to form a single storage surface.