Safety cabinets for flammable, BSC classic
Leverantör: LACONT UMWELTTECHNIK
Certifikat
Om denna produkt
Fire protection container is suitable for indoor and outdoor use. Fire resistant: 90 m.
- Steel sump as per WHG with grid cover
- With fire chamber test
- Doors close automatically if fire occurs
- Lifting attachments for cranes
Outer body painted in RAL 7038, agate-grey and inner body painted in RAL 9001, creme-white.