Safety Bottle Container
Supplier: Merck
Certificates
About this item
These bucket-style bottle carriers are essential safety equipment for transportation of hazardous chemicals.
- Versions for 2,5 L and 4 L glass bottles
- Additional bottle protection by a special developed carrier with foam inlay
- Solvent-resistant materials
- Leak-proof top cover