5-Bromophthalide
Catalog # MOLEM22752085
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:213.03 g/mol
EINECS:447-190-5
CAS Number:64169-34-2
Formula:C₈H₅BrO₂
MDL Number:MFCD01797360