Order Entry
Sweden
ContactUsLinkComponent
Chromium(III) chloride hexahydrate 98-101%, GPR RECTAPUR®
Irritant
Chromium(III) chloride hexahydrate 98-101%, GPR RECTAPUR®
Katalog # 84839.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Chromium(III) chloride hexahydrate 98-101%, GPR RECTAPUR®
Katalog # 84839.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    98 - 101 %
  • Insolubility in water:
    Max. 100 ppm
  • Substances not precipitated by NH4OH:
    Max. 0.2 %
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 100 ppm

Specification Test Results

About this item

 :
 :266.45 g/mol
Merck Index:13,02242
Melting Pt:85 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-038-3
UN:3260
ADR:8,III
 :GHS07
CAS Number:10060-12-5
Formula:CrCl₃·6H₂O
MDL Number:MFCD00149660