Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Personal Identification
ID Card Holders
ID-card holder, Kwintet
ID-card holder, Kwintet
ID-card holder, Kwintet
Catalog # KWIN100550940
Supplier:
FRISTADS KANSAS
Expand image
ID-card holder, Kwintet
Catalog # KWIN100550940
Supplier:
FRISTADS KANSAS
Specifications
Colour:
Black
Description:
ID-card holder, plastic, attached with clip or snap hook, pack of 10
Pk:
10
Specifications
About this item
Product Details
Documentation
Product Details
Documentation