He-Ne laser
Leverantör: DIDALAB
Certifikat
Om denna produkt
High quality professional gas laser, Class II
- Max Power:<1 mW
- Built-in tube power supply
- Divergence <2 mrad for unpolarised laser
Delivery: Supplied with rod Ø 10×130 mm for fitting to optical bench and mains power adapter