EDTA (ethylenediamine tetraacetic acid) 0.001786 mol/l in aqueous solution
Catalog # FFCHFF63421000
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:292.25 g/mol
Storage Temperature:Ambient
EINECS:200-449-4
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541