Order Entry
Sweden
ContactUsLinkComponent
Orthophosphoric acid 0.1 mol/l (0.3 N)
Orthophosphoric acid 0.1 mol/l (0.3 N)
Catalog # FFCHFF64325000
Supplier:  FF CHEMICALS
CAS Number:  
Orthophosphoric acid 0.1 mol/l (0.3 N)
Catalog # FFCHFF64325000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :98 g/mol
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340