Order Entry
Sweden
ContactUsLinkComponent
Ammonium acetate 10% (w/v) in aqueous solution
Ammonium acetate 10% (w/v) in aqueous solution
Catalog # FFCHFF62940500
Supplier:  FF CHEMICALS
CAS Number:  
undefined
Ammonium acetate 10% (w/v) in aqueous solution
Catalog # FFCHFF62940500
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :77.08 g/mol
Storage Temperature:Ambient
EINECS:211-162-9
CAS Number:631-61-8
Formula:H₃CCOONH₄
MDL Number:MFCD00013066